Läs mer
N-Methyl Viloxazine Hydrochloride, TRC
Produktkod. 30475108
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30475108 10 mg
1 options
Produktkod. 30475108 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33851110MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methyl Viloxazine Hydrochloride
CAS 35609-88-2
Molekylformel C14H21NO3 . HCl
Kvantitet 10 mg
Synonym 2-[(2-Ethoxyphenoxy)methyl]-4-methylmorpholine Hydrochloride, ICI 70098
InChI formel InChI=1S/C14H21NO3.ClH/c1-3-16-13-6-4-5-7-14(13)18-11-12-10-15(2)8-9-17-12;/h4-7,12H,3,8-11H2,1-2H3;1H
LEDER Cl.CCOc1ccccc1OCC2CN(C)CCO2
IUPAC-namn 2-[(2-ethoxyphenoxy)methyl]-4-methylmorpholine;hydrochloride
Molekylvikt (g/mol) 287.78
Formel vikt 287.1288