Läs mer
N-Methylacetamide-N-d1, CDN
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Methylacetamide-N-D |
| CAS | 3669-70-3 |
| Molekylformel | C3 D H6 N O |
| Kvantitet | 1 g |
| Synonym | Acetamide-N-d, N-methyl- (6 CI, 7 CI, 8 CI, 9 CI), N-Deutero-N-methylacetamide, N-Methylacetamide-N-d1, N-Methylacetamide-d, N-Methylacetamide-N-D |
| InChI formel | InChI=1 S/C3H7NO/c1-3(5)4-2/h1-2H3,(H,4,5)/i/hD |
| LEDER | [2 H]N(C)C(=O)C |
| IUPAC-namn | N-deuterio-N-methylacetamide |
| Molekylvikt (g/mol) | 74.1 |
| Formel vikt | 74.059 g/mol |
| Visa mer |