Läs mer
N-Methylacetoacetamide (70% in aqueous solution), TRC
Produktkod. 30483390
Change view
Kvantitet:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483390 25 g
30472924 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30483390 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM29322525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20306-75-6
Molekylformel C5H9NO2
Molekylvikt (g/mol) 115.13
Synonym N-Methyl-3-oxobutanamide, 2-Acetyl-N-methylacetamide
IUPAC-namn N-methyl-3-oxobutanamide
LEDER CC(CC(NC)=O)=O
Specifikationer
Kemiskt namn eller material N-Methylacetoacetamide
Kvantitet 25 g
InChI formel InChI=1S/C5H9NO2/c1-4(7)3-5(8)6-2/h3H2,1-2H3,(H,6,8)
Formel vikt 115.0633
Koncentration eller sammansättning (efter analyt eller komponenter) N-Methylacetoacetamide 70 %