Läs mer
N-Methylaniline Hydrochloride, TRC
Produktkod. 30484031
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30484031 10 g
30470487 1 g
30480519 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30484031 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28830810G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2739-12-0
Molekylformel C7H9N . HCl
Molekylvikt (g/mol) 143.61
Synonym N-Methylanilinium Chloride, N-Methylbenzenamine Hydrochloride, Anilinomethane Hydrochloride
IUPAC-namn N-methylaniline;hydrochloride
LEDER Cl.CNc1ccccc1
Specifikationer
Kemiskt namn eller material N-Methylaniline Hydrochloride
Kvantitet 10 g
InChI formel InChI=1S/C7H9N.ClH/c1-8-7-5-3-2-4-6-7;/h2-6,8H,1H3;1H
Formel vikt 143.0502