Läs mer
N-Methylaniline Trifluoroacetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Methylaniline Trifluoroacetate |
| CAS | 29885-95-8 |
| Molekylformel | C7H9N . C2HF3O2 |
| Kvantitet | 2.5 g |
| Synonym | Benzenamine','N-methyl-','2,2,2-trifluoroacetate (1:1) (ACI), Acetic acid','trifluoro-','compd. with N-methylaniline (1:1) (8CI), Acetic acid','trifluoro-','compd. with N-methylbenzenamine (1:1) (ZCI), Aniline','N-methyl-','trifluoroacetate (8CI), Benzenamine','N-methyl-','trifluoroacetate (9CI), N-Methylaniline trifluoroacetic acid salt, N-Methylanilinium trifluoroacetate, N-Methylaniline Trifluoroacetate |
| InChI formel | InChI=1S/C7H9N.C2HF3O2/c1-8-7-5-3-2-4-6-7;3-2(4,5)1(6)7/h2-6,8H,1H3;(H,6,7) |
| LEDER | CNc1ccccc1.OC(=O)C(F)(F)F |
| IUPAC-namn | N-methylaniline;2,2,2-trifluoroacetic acid |
| Molekylvikt (g/mol) | 221.18 |
| Formel vikt | 221.0664 |