Läs mer
N-Methylbut-2-enoate Ethyl Ester D-(-)Phenylglycine Potassium Salt, TRC
Produktkod. 30475453
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475453 5 g
30468667 1 g
30475645 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475453 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2951355G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 961-69-3
Molekylformel C14H16KNO4
Molekylvikt (g/mol) 301.38
Synonym (R)-alpha-[(3-Ethoxy-1-methyl-3-oxo-1-propenyl)amino]benzeneacetic Acid Monopotassium Salt, (alphaR)-alpha-[(3-Ethoxy-1-methyl-3-oxo-1-propenyl)amino]benzeneacetic Monopotassium Salt, D-(-)-3-[(alpha-Carboxybenzyl)amino]crotonic Acid 1-Ethyl Ester Monopotassium Salt, Potassium D-(1-Ethoxycarbonylpropen-2-yl)-alpha-aminophenylacetate
IUPAC-namn potassium;(2R)-2-[(4-ethoxy-4-oxobut-2-en-2-yl)amino]-2-phenylacetate
LEDER [K+].CCOC(=O)C=C(C)N[C@@H](C(=O)[O-])c1ccccc1
Specifikationer
Kemiskt namn eller material N-Methylbut-2-enoate Ethyl Ester D-(-)Phenylglycine Potassium Salt
Kvantitet 5 g
InChI formel InChI=1S/C14H17NO4.K/c1-3-19-12(16)9-10(2)15-13(14(17)18)11-7-5-4-6-8-11;/h4-9,13,15H,3H2,1-2H3,(H,17,18);/q;+1/p-1/t13-;/m1./s1
Formel vikt 301.0716