Läs mer
N-Methylcytisine, TRC
Produktkod. 17964195
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17964195 10 mg
17930465 50 mg
17991275 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17964195 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM29468010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 486-86-2
Molekylformel C12 H16 N2 O
Molekylvikt (g/mol) 204.27
Synonym (-)-N-Methylcytisine, 1,5-Methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, 1,2,3,4,5,6-hexahydro-3-methyl-, (1R,5S)-, 1,5-Methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, 1,2,3,4,5,6-hexahydro-3-methyl-, (1R)-, Cytisine, 12-methyl- (8CI), Cytisine, N-methyl- (7CI), Cytisine, methyl- (6CI), (-)-N-Methylcytisine, (1R,5S)-1,2,3,4,5,6-Hexahydro-3-methyl-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, Caulophyllin, Caulophylline, N-Methylcytisine
LEDER CN1C[C@@H]2C[C@H](C1)C3=CC=CC(=O)N3C2
Specifikationer
Kemiskt namn eller material N-Methylcytisine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C12H16N2O/c1-13-6-9-5-10(8-13)11-3-2-4-12(15)14(11)7-9/h2-4,9-10H,5-8H2,1H3/t9-,10+/m0/s1
Formel vikt 204.1263
Procent renhet >95%