Läs mer
N-Methyldeoxynojirimycin, TRC
Produktkod. 30479969
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479969 25 mg
30475099 5 mg
30468335 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479969 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM29700025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 69567-10-8
Molekylformel C7 H15 N O4
Molekylvikt (g/mol) 177.2
Synonym N-Methyl-1-Deoxynojirimycin
IUPAC-namn (2R,3R,4R,5S)-2-(hydroxymethyl)-1-methylpiperidine-3,4,5-triol
LEDER CN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO
Specifikationer
Kemiskt namn eller material N-Methyldeoxynojirimycin
Kvantitet 25 mg
InChI formel InChI=1S/C7H15NO4/c1-8-2-5(10)7(12)6(11)4(8)3-9/h4-7,9-12H,2-3H2,1H3/t4-,5+,6-,7-/m1/s1
Formel vikt 177.1001