Läs mer
N-Methylisoamylamine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Methylisoamylamine Hydrochloride |
| CAS | 2419-59-2 |
| Molekylformel | C6H15N .HCl |
| Kvantitet | 1 g |
| Synonym | Methylisopentylamine Hydrochloride, N-Isoamyl-N-methylamine Hydrochloride, N-Methyl-3-methylbutanamine Hydrochloride, N-Methyl-3-methylbutylamine Hydrochloride, N-Methyl-N-(3-methylbutyl)amine Hydrochloride, N-Methylisoamylamine Hydrochloride, N-Methylisopentylamine Hydrochloride |
| InChI formel | InChI=1S/C6H15N.ClH/c1-6(2)4-5-7-3;/h6-7H,4-5H2,1-3H3;1H |
| LEDER | Cl.CNCCC(C)C |
| IUPAC-namn | N,3-dimethylbutan-1-amine;hydrochloride |
| Molekylvikt (g/mol) | 137.65 |
| Formel vikt | 137.0971 |