missing translation for 'onlineSavingsMsg'
Läs mer

N-Methylmethanesulfonamide Sodium Salt, TRC

Produktkod. 30469633
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30469633 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30469633 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3231901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-Methylmethanesulfonamide Sodium Salt
CAS 41881-75-8
Molekylformel C2H6NNaO2S
Kvantitet 1 g
InChI formel InChI=1S/C2H6NO2S.Na/c1-3-6(2,4)5;/h1-2H3;/q-1;+1
LEDER CS(=O)([N-]C)=O.[Na+]
IUPAC-namn sodium methyl(methylsulfonyl)amide
Molekylvikt (g/mol) 131.11
Formel vikt 131
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.