Läs mer
N-Methylperfluorooctanesulfonamide, TRC
Produktkod. 30472186
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472186 25 mg
30471725 50 mg
30476160 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30472186 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32734025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 31506-32-8
Molekylformel C9 H4 F17 N O2 S
Molekylvikt (g/mol) 513.17
Synonym 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N-methyl-1-octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N-methyloctane-1-sulfonamide, AI 3-29758, Fluorad FX 12, N-MeFOSA, N-Methyl perfluorooctanesulphonamide, N-Methylheptadecafluorooctanesulfonamide, N-Methylperfluorooctanesulfonamide, Perfluoro-N-methyloctanesulfonamide, MeFOSA
IUPAC-namn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-methyloctane-1-sulfonamide
LEDER CNS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material N-Methylperfluorooctanesulfonamide
Kvantitet 25 mg
InChI formel InChI=1S/C9H4F17NO2S/c1-27-30(28,29)9(25,26)7(20,21)5(16,17)3(12,13)2(10,11)4(14,15)6(18,19)8(22,23)24/h27H,1H3
Formel vikt 512.9691