Läs mer
N-Methylsuccinimide, TRC
Produktkod. 30475543
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30475543 250 mg
1 options
Produktkod. 30475543 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM329425250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methylsuccinimide
CAS 1121-07-9
Molekylformel C5H7NO2
Kvantitet 250 mg
Synonym 1-Methyl-2,5-pyrrolidinedione, N-Methyl-2,5-pyrrolidinedione, NSC 11324
InChI formel InChI=1S/C5H7NO2/c1-6-4(7)2-3-5(6)8/h2-3H2,1H3
LEDER CN1C(=O)CCC1=O
IUPAC-namn 1-methylpyrrolidine-2,5-dione
Molekylvikt (g/mol) 113.11
Formel vikt 113.0477