Läs mer
N-(Methylsulfonyl)methanesulfonamide, TRC
Produktkod. 30477131
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30477131 250 mg
1 options
Produktkod. 30477131 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM645890250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(Methylsulfonyl)methanesulfonamide
CAS 5347-82-0
Molekylformel C2 H7 N O4 S2
Kvantitet 250 mg
Synonym Bis(methanesulfonyl)amine
InChI formel InChI=1S/C2H7NO4S2/c1-8(4,5)3-9(2,6)7/h3H,1-2H3
LEDER CS(=O)(=O)NS(=O)(=O)C
IUPAC-namn N-methylsulfonylmethanesulfonamide
Molekylvikt (g/mol) 173.211
Formel vikt 172.982