Läs mer
N-Methylthiophene-2-carboxamide, Mikromol™
Produktkod. 30463583
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30463583 4 x 25 mg
17982236 100 mg
17941836 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30463583 Leverantör Mikromol Leverantörsnummer MM0247.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Methylthiophene-2-carboxamide, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methylthiophene-2-carboxamide
CAS 39880-77-8
Analyt- eller komponentnamn N-Methylthiophene-2-carboxamide
Frakt skick Room Temperature
Molekylformel C6 H7 N O S
Certifikat och efterlevnad ISO 17025
Synonym N-Methylthiophene-2-carboxamide, Tenoxicam Imp. C (EP), Tenoxicam Impurity C
InChI formel InChI=1S/C6H7NOS/c1-7-6(8)5-3-2-4-9-5/h2-4H,1H3,(H,7,8)
Lösningstyp Neat
LEDER CNC(=O)c1cccs1
IUPAC-namn N-methylthiophene-2-carboxamide
Molekylvikt (g/mol) 141.19
Kvantitet 4 x 25 mg
Formel vikt 141.0248 g/mol