Läs mer
N-Methylvancomycin Di-trifluoroacetic Acid Salt (>85%), TRC
Produktkod. 30180325
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180325 10 mg
30179987 1 mg
30180120 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180325 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM34851510MG
 Begränsad artikel
Beräknad leverans: 26-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
Synonym 22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine, Vancomycin Derivative Di-trifluoroacetic Acid Salt∣56-Methylvancomycin Di-trifluoroacetic Acid Salt;∣Antibiotic M 43D Di-trifluoroacetic Acid Salt∣(3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-(((2S,3R,4S,5S,6R)-3-(((2S,4S,5S,6S)-4-amino-5-hydroxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-43,45-dichloro-6-((R)-2-(dimethylamino)-4-methylpentanamido)-7,22,28,30,32-pentahydroxy-2,5,24,38,39-pentaoxo-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-1H,22H-23,36-(epiminomethano)-8,11:18,21-dietheno-13,16:31,35-di(metheno)benzo[n][1]oxa[6,9]diazacyclohexadecino[4,5-d][1]oxa[7,17]diazacyclotetracosine-26-carboxylic Acid Di-trifluoroacetic Acid Salt
Specifikationer
Formatera Neat
Molekylvikt (g/mol) 1691.33
Produkttyp API