Läs mer
N,N’-Bis(2,3-dihydroxypropyl)-5-nitro-1,3-benzenedicarboxamide, TRC
Produktkod. 30136131
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136131 5 g
17938604 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136131 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4266005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 76820-34-3
Molekylformel C14 H19 N3 O8
Molekylvikt (g/mol) 357.32
Synonym N,N'-bis(2,3-dihydroxypropyl)-5-nitro-1,3-benzenedicarboxamide
IUPAC-namn 1-N,3-N-bis(2,3-dihydroxypropyl)-5-nitrobenzene-1,3-dicarboxamide
LEDER OCC(O)CNC(=O)c1cc(cc(c1)[N+](=O)[O-])C(=O)NCC(O)CO
Specifikationer
Kemiskt namn eller material N,N'-bis(2,3-dihydroxypropyl)-5-nitro-1,3-benzenedicarboxamide
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C14H19N3O8/c18-6-11(20)4-15-13(22)8-1-9(3-10(2-8)17(24)25)14(23)16-5-12(21)7-19/h1-3,11-12,18-21H,4-7H2,(H,15,22)(H,16,23)
Formel vikt 357.1172
Procent renhet >95%