missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(acryloyl)cystamine, Electrophoresis Reagent, 98%, Thermo Scientific Chemicals

Catalog Number p-7057560
Click to view available options
Quantity:
1 g
5 g
25 g
60984-57-8
C10H16N2O2S2
260.37
MFCD00036225
DJVKJGIZQFBFGS-UHFFFAOYSA-N
n,n'-bis acryloyl cystamine, n,n'-bisacrylylcystamine, 2-propenamide, n,n'-dithiodi-2,1-ethanediyl bis, 2,2'-bisacrylamino diethyldisulfide, bis-acrylylcystamine: bac, n,n'-dithiodi-2,1-ethanediyl bis acrylamide, n-2-2-prop-2-enamido ethyl disulfanyl ethyl prop-2-enamide, n-2-2-prop-2-enoylamino ethyldisulfanyl ethyl prop-2-enamide, acmc-20ak9p, n,n-bis-acrylylcystamine
100602
N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide
C=CC(=O)NCCSSCCNC(=O)C=C
60984-57-8
C10H16N2O2S2
260.37
MFCD00036225
DJVKJGIZQFBFGS-UHFFFAOYSA-N
n,n'-bis acryloyl cystamine, n,n'-bisacrylylcystamine, 2-propenamide, n,n'-dithiodi-2,1-ethanediyl bis, 2,2'-bisacrylamino diethyldisulfide, bis-acrylylcystamine: bac, n,n'-dithiodi-2,1-ethanediyl bis acrylamide, n-2-2-prop-2-enamido ethyl disulfanyl ethyl prop-2-enamide, n-2-2-prop-2-enoylamino ethyldisulfanyl ethyl prop-2-enamide, acmc-20ak9p, n,n-bis-acrylylcystamine
100602
N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide
C=CC(=O)NCCSSCCNC(=O)C=C

BAC is used as a reversible cross-linker for polyacrylamide gels, to simplify isolation of macromolecules separated on the gels. The disulfide linkage can be broken with a suitable reducing agent. A 1-g slice of a 4% (12:1) gel will dissolve in 5 mL of 100 mM DTT in TBE (pH 8.3) in less than 30 min. The pH of 8.3 is optimum for rapid dissolution of the gel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
BAC is used as a reversible cross-linker for polyacrylamide gels, to simplify isolation of macromolecules separated on the gels. The disulfide linkage can be broken with a suitable reducing agent. A 1-g slice of a 4% (12:1) gel will dissolve in 5 mL of 100 mM DTT in TBE (pH 8.3) in less than 30 min. The pH of 8.3 is optimum for rapid dissolution of the gel.

Solubility
Soluble at 50 mg/ml in acetic acid

Notes
Store at -20°C. Incompatible with heat, oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 123°C to 124°C
Color White
Quantity 1 g
Solubility Information Soluble at 50mg/mL in acetic acid
Formula Weight 260.37
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N,N'-Bis(acryloyl)cystamine, Electrophoresis Reagent

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'