Läs mer
N,N’-Bis(benzyloxycarbonyl)-L-lysine 4-Nitrobenzyl Ester, TRC
Produktkod. 30341141
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30341141 250 mg
1 options
Produktkod. 30341141 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB410910250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-Bis(benzyloxycarbonyl)-L-lysine 4-Nitrobenzyl Ester
CAS 1331899-68-3
Molekylformel C29 H31 N3 O8
Kvantitet 250 mg
Synonym N,N'-Bis(Benzyloxycarbonyl)-L-Lysine 4-Nitrobenzyl Ester
InChI formel InChI=1S/C29H31N3O8/c33-27(38-19-24-14-16-25(17-15-24)32(36)37)26(31-29(35)40-21-23-11-5-2-6-12-23)13-7-8-18-30-28(34)39-20-22-9-3-1-4-10-22/h1-6,9-12,14-17,26H,7-8,13,18-21H2,(H,30,34)(H,31,35)/t26-/m0/s1
LEDER [O-][N+](=O)c1ccc(COC(=O)[C@H](CCCCNC(=O)OCc2ccccc2)NC(=O)OCc3ccccc3)cc1
IUPAC-namn (4-nitrophenyl)methyl (2S)-2,6-bis(phenylmethoxycarbonylamino)hexanoate
Molekylvikt (g/mol) 549.57
Formel vikt 549.2111