Läs mer
N,N-bis-(Carboxymethyl)-L-glutamic Acid Tetrasodiumn Salt (40% in water), TRC
Produktkod. 30010089
Change view
Kvantitet:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010089 10 g
30007571 5 g
30006159 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010089 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC18115810G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51981-21-6
Molekylformel C9H9NNa4O8
Molekylvikt (g/mol) 351.13
IUPAC-namn tetrasodium;(2S)-2-[bis(carboxylatomethyl)amino]pentanedioate
LEDER [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CC[C@H](N(CC(=O)[O-])CC(=O)[O-])C(=O)[O-]
Specifikationer
Kemiskt namn eller material N,N-bis-(Carboxymethyl)-L-glutamic Acid Tetrasodiumn Salt
Kvantitet 10 g
InChI formel InChI=1S/C9H13NO8.4Na/c11-6(12)2-1-5(9(17)18)10(3-7(13)14)4-8(15)16;;;;/h5H,1-4H2,(H,11,12)(H,13,14)(H,15,16)(H,17,18);;;;/q;4*+1/p-4/t5-;;;;/m0..../s1
Formel vikt 350.9919
Koncentration eller sammansättning (efter analyt eller komponenter) N,N-bis-(Carboxymethyl)-L-glutamic Acid Tetrasodiumn Salt 40%