Läs mer
N,N'-bis(D-Glucopyranosyl)-4,4’bis(diphenylamino)methane (Mixture of Diastereomers), TRC
Produktkod. 30365397
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30365397 100 mg
1 options
Produktkod. 30365397 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD271570100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-bis(D-Glucopyranosyl)-4,4'bis(diphenylamino)methane (Mixture of Diastereomers)
CAS 30796-64-6
Molekylformel C25 H34 N2 O10
Kvantitet 100 mg
Synonym Dextrose Adduct 2
InChI formel InChI=1S/C25H34N2O10/c28-10-16-18(30)20(32)22(34)24(36-16)26-14-5-1-12(2-6-14)9-13-3-7-15(8-4-13)27-25-23(35)21(33)19(31)17(11-29)37-25/h1-8,16-35H,9-11H2/t16-,17-,18-,19-,20+,21+,22-,23-,24?,25?/m1/s1
LEDER OC[C@H]1OC(Nc2ccc(Cc3ccc(NC4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)cc3)cc2)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC-namn (2R,3S,4S,5R)-2-(hydroxymethyl)-6-[4-[[4-[[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]amino]phenyl]methyl]anilino]oxane-3,4,5-triol
Molekylvikt (g/mol) 522.545
Formel vikt 522.221