Läs mer
N,N'-Bis(trifluoroacetyl)-L-Cystine Dimethyl Ester, TRC
Produktkod. 30342926
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30342926 5 g
1 options
Produktkod. 30342926 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB5855155G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-Bis(trifluoroacetyl)-L-Cystine Dimethyl Ester
CAS 1492-83-7
Molekylformel C12H14F6N2O6S2
Kvantitet 5 g
Synonym L-N,N'-bis(trifluoroacetyl)cystine Dimethyl Ester, N,N'-Bis(2,2,2-trifluoroacetyl)-L-cystine 1,1’-Dimethyl Ester
InChI formel InChI=1S/C12H14F6N2O6S2/c1-25-7(21)5(19-9(23)11(13,14)15)3-27-28-4-6(8(22)26-2)20-10(24)12(16,17)18/h5-6H,3-4H2,1-2H3,(H,19,23)(H,20,24)/t5-,6-/m0/s1
LEDER COC(=O)[C@H](CSSC[C@H](NC(=O)C(F)(F)F)C(=O)OC)NC(=O)C(F)(F)F
IUPAC-namn methyl (2R)-3-[[(2R)-3-methoxy-3-oxo-2-[(2,2,2-trifluoroacetyl)amino]propyl]disulfanyl]-2-[(2,2,2-trifluoroacetyl)amino]propanoate
Molekylvikt (g/mol) 460.4
Formel vikt 460.0197