Läs mer
N-(n-Butyl)-phosphoramidic Dichloride, TRC
Produktkod. 30345379
Change view
Kvantitet:
50 g
Produktkod. Kvantitet
30345379 50 g
1 options
Produktkod. 30345379 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB69364050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(n-Butyl)-phosphoramidic Dichloride
CAS 90206-85-2
Molekylformel C14 H25 N O11
Kvantitet 50 g
Synonym Butyl-phosphoramidic dichloride
InChI formel InChI=1S/C14H25NO11/c1-4(18)15-7-9(20)8(19)5(2-16)25-14(7)26-12-6(3-17)24-13(23)11(22)10(12)21/h5-14,16-17,19-23H,2-3H2,1H3,(H,15,18)/t5?,6?,7?,8-,9?,10?,11?,12-,13+,14-/m0/s1
LEDER CC(=O)NC1C(O)[C@@H](O)C(CO)O[C@H]1O[C@@H]2C(O)C(O)[C@H](O)OC2CO
IUPAC-namn N-[(2S,5R)-4,5-dihydroxy-6-(hydroxymethyl)-2-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]acetamide
Molekylvikt (g/mol) 383.348
Formel vikt 383.143