Läs mer
N,N'-Carbonylbis[L-glutamic Acid], TRC
Produktkod. 30352609
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30352609 25 mg
1 options
Produktkod. 30352609 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17730825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-Carbonylbis[L-glutamic Acid]
CAS 302941-52-2
Molekylformel C11 H16 N2 O9
Kvantitet 25 mg
Synonym Carglumic Acid Dimer
InChI formel InChI=1S/C11H16N2O9/c14-7(15)3-1-5(9(18)19)12-11(22)13-6(10(20)21)2-4-8(16)17/h5-6H,1-4H2,(H,14,15)(H,16,17)(H,18,19)(H,20,21)(H2,12,13,22)/t5-,6-/m0/s1
LEDER OC(=O)CC[C@H](NC(=O)N[C@@H](CCC(=O)O)C(=O)O)C(=O)O
IUPAC-namn (2S)-2-[[(1S)-1,3-dicarboxypropyl]carbamoylamino]pentanedioic acid
Molekylvikt (g/mol) 320.253
Formel vikt 320.086