Läs mer
N,N’-Desethylene-N-Desmethyl Levofloxacin Hydrochloride Hydrochloride, TRC
Produktkod. 30006669
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006669 50 mg
30005448 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006669 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28934050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1797099-76-3
Molekylformel C15 H16 F N3 O4 . Cl H
Molekylvikt (g/mol) 357.7646
Synonym 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 10-[(2-aminoethyl)amino]-9-fluoro-2,3-dihydro-3-methyl-7-oxo-, hydrochloride (1:1), (3S)-, N,N'-Desethylene-N-desmethyl Levofloxacin hydrochloride, Levofloxacin diamine hydrochloride, N,N'-Desethylene-N-desmethyllevofloxacin hydrochloride, (3S)-10-[(2-Aminoethyl)amino]-9-fluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid hydrochloride
LEDER Cl.C[C@H]1COc2c(NCCN)c(F)cc3C(=O)C(=CN1c23)C(=O)O
Specifikationer
Kemiskt namn eller material N,N'-Desethylene-N-desmethyllevofloxacin Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C15H16FN3O4.ClH/c1-7-6-23-14-11(18-3-2-17)10(16)4-8-12(14)19(7)5-9(13(8)20)15(21)22;/h4-5,7,18H,2-3,6,17H2,1H3,(H,21,22);1H/t7-;/m0./s1
Formel vikt 357.0892