Läs mer
N,N'-Desethylene-N,N'-diformyl Levofloxacin, TRC
Produktkod. 30361089
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30361089 25 mg
1 options
Produktkod. 30361089 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28933025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-Desethylene-N,N'-diformyl Levofloxacin
CAS 151377-74-1
Molekylformel C18H18FN3O6
Kvantitet 25 mg
Synonym (S)-9-Fluoro-10-[formyl[2-(formylmethylamino)ethyl]amino]-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid, Levofloxacin Desethylene Diformyl Impurity
InChI formel InChI=1S/C18H18FN3O6/c1-10-7-28-17-14-11(16(25)12(18(26)27)6-22(10)14)5-13(19)15(17)21(9-24)4-3-20(2)8-23/h5-6,8-10H,3-4,7H2,1-2H3,(H,26,27)/t10-/m0/s1
LEDER C[C@H]1COC2=C3N1C=C(C(=O)C3=CC(=C2N(CCN(C)C=O)C=O)F)C(=O)O
IUPAC-namn (S)-9-Fluoro-3-methyl-10-(N-(2-(N-methylformamido)ethyl)formamido)-7-oxo-2,3-dihydro-7H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic Acid
Molekylvikt (g/mol) 391.35
Formel vikt 391.12