Läs mer
N,N’-Desethylene Ofloxacin Hydrochloride, TRC
Produktkod. 30134462
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134462 500 mg
30007043 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134462 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD289365500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1797983-25-5
Molekylformel C16H19ClFN3O4
Molekylvikt (g/mol) 371.79
Synonym 9-Fluoro-2,3-dihydro-3-methyl-10-[[2-(methylamino)ethyl]amino]-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid Hydrochloride, Ofloxacin Desethylene Impurity
LEDER Cl.CNCCNc1c(F)cc2C(=O)C(=CN3C(C)COc1c23)C(=O)O
Specifikationer
Kemiskt namn eller material N,N'-Desethylene Ofloxacin Hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C16H18FN3O4.ClH/c1-8-7-24-15-12(19-4-3-18-2)11(17)5-9-13(15)20(8)6-10(14(9)21)16(22)23;/h5-6,8,18-19H,3-4,7H2,1-2H3,(H,22,23);1H
Formel vikt 371.1048