Läs mer
N,N-Diamylnitrosamine, TRC
Produktkod. 30357331
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30357331 25 mg
1 options
Produktkod. 30357331 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41686425MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N-Diamylnitrosamine
CAS 13256-06-9
Molekylformel C10 H22 N2 O
Kvantitet 25 mg
Synonym 1-Pentanamine, N-nitroso-N-pentyl- (9CI, ACI), Dipentylamine, N-nitroso- (6CI, 7CI, 8CI), N-Nitroso-N-pentyl-1-pentanamine (ACI), Diamylnitrosamine, Dipentylnitrosamine, Dipentylnitrosoamine, N,N-Dipentylnitrosamine, N-Nitrosodi-n-amylamine, N-Nitrosodi-n-pentylamine, N-Nitrosodiamylamine, N-Nitrosodipentylamine, NSC 73601
InChI formel InChI=1S/C10H22N2O/c1-3-5-7-9-12(11-13)10-8-6-4-2/h3-10H2,1-2H3
LEDER CCCCCN(CCCCC)N=O
IUPAC-namn N,N-dipentylnitrous amide
Molekylvikt (g/mol) 186.29
Formel vikt 186.1732