missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Dibenzylethylenediamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
N,N'-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N′-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
N,N′-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Specifications
Melting Point | 23°C to 25°C |
Density | 1.025 |
Boiling Point | 184°C to 186°C (2 mmHg) |
Flash Point | >110°C (230°F) |
Linear Formula | C6H5CH2NHCH2CH2NHCH2C6H5 |
Refractive Index | 1.565 |
Quantity | 25 g |
UN Number | UN2735 |
Beilstein | 786668 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'