Läs mer
N,N-Dibenzylformamide, TRC
Produktkod. 30350048
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30350048 1 g
1 options
Produktkod. 30350048 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4171001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N-Dibenzylformamide
CAS 5464-77-7
Molekylformel C15H15NO
Kvantitet 1 g
Synonym N,N-Bis(phenylmethyl)formamide, Dibenzylformamide, NSC 5982
InChI formel InChI=1S/C15H15NO/c17-13-16(11-14-7-3-1-4-8-14)12-15-9-5-2-6-10-15/h1-10,13H,11-12H2
LEDER O=CN(Cc1ccccc1)Cc2ccccc2
IUPAC-namn N,N-dibenzylformamide
Molekylvikt (g/mol) 225.29
Formel vikt 225.1154