Läs mer
N,N-Dicyclohexyl-2-benzothiazolsulfene Amide, TRC
Produktkod. 30375153
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30375153 500 mg
1 options
Produktkod. 30375153 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD439458500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N-Dicyclohexyl-2-benzothiazolsulfene Amide
CAS 4979-32-2
Anteckningar om renhetsgrad HPLC
Molekylformel C19 H26 N2 S2
Kvantitet 500 mg
Synonym N,N-Dicyclohexyl-2-benzothiazolesulfenamide (ACI), 2-Benzothiazolyl-N,N-dicyclohexylsulfenamide, Accel DZ-G, Accelerator DZ, DCBS, DTs, M 181, Meramid DCH, Mercure DCBS, N,N-Dicyclohexyl-1,3-benzothiazole-2-sulfenamide, N,N-Dicyclohexyl-2-benzothiazolylsulfenamide, N,N-Dicyclohexyl-2-mercaptobenzothiazole sulfenamide, Nocceler DZ, Nocceler DZ-G, Rhodifax 30, SA-DTs, Sanceler DZ, Sanceler DZ-G, Santocure DCBS, Soxinol DZ, Sulfenamide DTs, Vulkacit DZ
InChI formel InChI=1S/C19H26N2S2/c1-3-9-15(10-4-1)21(16-11-5-2-6-12-16)23-19-20-17-13-7-8-14-18(17)22-19/h7-8,13-16H,1-6,9-12H2
LEDER C1CCC(CC1)N(Sc2nc3ccccc3s2)C4CCCCC4
IUPAC-namn N-(1,3-benzothiazol-2-ylsulfanyl)-N-cyclohexyl-cyclohexanamine
Molekylvikt (g/mol) 346.55
Formel vikt 346.1537