Läs mer
(N,N-didemethyl) 1-Hydroxy Sibutramine Hydrochloride, TRC
Produktkod. 30133074
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133074 50 mg
30006197 25 mg
30008578 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133074 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29838550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 186521-88-0
Molekylformel C15H23Cl2NO
Molekylvikt (g/mol) 304.26
Synonym δ-Amino-1-(4-chlorophenyl)-β-methylcyclobutanebutanol Hydrocholride
LEDER CC(CO)CC(N)C1(CCC1)C2=CC=C(Cl)C=C2.Cl
Specifikationer
Kemiskt namn eller material (N,N-didemethyl) 1-Hydroxy Sibutramine Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C15H22ClNO.ClH/c1-11(10-18)9-14(17)15(7-2-8-15)12-3-5-13(16)6-4-12;/h3-6,11,14,18H,2,7-10,17H2,1H3;1H
Formel vikt 303.1157
Procent renhet >95%