Läs mer
N,N-Didemethylorphenadrine Hydrochloride, TRC
Produktkod. 30134819
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134819 250 mg
30008330 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134819 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD439700250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17752-32-8
Molekylformel C16 H19 N O . Cl H
Molekylvikt (g/mol) 277.79
Synonym (RS)-2-[(2-Methylphenyl)phenylmethoxy]ethanamine Hydrochloride, Orphenadrine Hydrochloride Imp. C (EP) as Hydrochloride, Ethanamine, 2-[(2-methylphenyl)phenylmethoxy]-, hydrochloride (1:1), Ethanamine, 2-[(2-methylphenyl)phenylmethoxy]-, hydrochloride (9CI), Ethylamine, 2-[(o-methyl-α-phenylbenzyl)oxy]-, hydrochloride (8CI), (o-Methyl-α-phenylbenzyloxy)ethylamine, hydrochloride, N,N-Didemethylorphenadrine hydrochloride
IUPAC-namn 2-[(2-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride
LEDER Cl.Cc1ccccc1C(OCCN)c2ccccc2
Specifikationer
Kemiskt namn eller material (RS)-2-[(2-Methylphenyl)phenylmethoxy]ethanamine Hydrochloride
Kvantitet 250 mg
InChI formel InChI=1S/C16H19NO.ClH/c1-13-7-5-6-10-15(13)16(18-12-11-17)14-8-3-2-4-9-14;/h2-10,16H,11-12,17H2,1H3;1H
Formel vikt 277.1233