Läs mer
N,N-Diethylbenzamide, TRC
Produktkod. 30373902
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30373902 5 g
1 options
Produktkod. 30373902 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4440805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N-Diethylbenzamide
CAS 1696-17-9
Molekylformel C11H15NO
Kvantitet 5 g
Synonym Benzoic Acid N,N-Diethylamide, Benzoic Acid Diethylamide, Benzoyldiethylamine, NSC 16060, R 2, R 2 (Insect Repellent), Rebemid, Rebemide
InChI formel InChI=1S/C11H15NO/c1-3-12(4-2)11(13)10-8-6-5-7-9-10/h5-9H,3-4H2,1-2H3
LEDER CCN(CC)C(=O)c1ccccc1
Molekylvikt (g/mol) 177.24
Formel vikt 177.1154