missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diformylacetamide, 80%, Thermo Scientific Chemicals

Catalog Number p-3523503
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
26944-31-0
C4H5NO3
115.09
MFCD00015916
ZAHBLOGBMDVRDK-UHFFFAOYSA-N
n,n-diformylacetamid
8063338
N,N-diformylacetamide
CC(=O)N(C=O)C=O
This item is not returnable. View return policy
26944-31-0
C4H5NO3
115.09
MFCD00015916
ZAHBLOGBMDVRDK-UHFFFAOYSA-N
n,n-diformylacetamid
8063338
N,N-diformylacetamide
CC(=O)N(C=O)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 75.0
CAS Max % 100.0
Color Colorless
Boiling Point 70°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 75% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(O)N(COH)2
Quantity 1 g
Formula Weight 115.09
Percent Purity 80%
Physical Form Liquid
Chemical Name or Material N, N-Diformylacetamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'