Läs mer
N,N-Dimethyl-2-chloroacetoacetamide, TRC
Produktkod. 30370614
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30370614 100 mg
1 options
Produktkod. 30370614 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD470726100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N-Dimethyl-2-chloroacetoacetamide
CAS 5810-11-7
Molekylformel C6H10ClNO2
Kvantitet 100 mg
Synonym 2-Chloro-N,N-dimethyl-3-oxobutanamide, 2-Chloro-N,N-dimethylacetoacetamide, 2-Chloroacetoacetic Acid N,N-dimethylamide, N,N-Dimethyl-2-chloroacetoacetamide
InChI formel InChI=1S/C6H10ClNO2/c1-4(9)5(7)6(10)8(2)3/h5H,1-3H3
LEDER CN(C)C(=O)C(Cl)C(=O)C
Molekylvikt (g/mol) 163.6
Formel vikt 163.04