Läs mer
N,N-Dimethyl-2-methylene-Beta-alanine, TRC
Produktkod. 30375530
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375530 100 mg
30372778 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30375530 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD474765100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5415-98-5
Molekylformel C6H11NO2
Molekylvikt (g/mol) 129.16
Synonym 2-[(Dimethylamino)methyl]-2-propenoic Acid, 2-[(Dimethylamino)methyl]acrylic Acid, NSC 11325
LEDER CN(C)CC(=C)C(=O)O
Specifikationer
Kemiskt namn eller material N,N-Dimethyl-2-methylene-beta-alanine
Kvantitet 100 mg
InChI formel InChI=1S/C6H11NO2/c1-5(6(8)9)4-7(2)3/h1,4H2,2-3H3,(H,8,9)
Formel vikt 129.079