Läs mer
N,N-Dimethyl-2-phenoxyethanamine, TRC
Produktkod. 30136826
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136826 500 mg
30132902 100 mg
30012731 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136826 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB412853500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13468-02-5
Molekylformel C10H15NO
Molekylvikt (g/mol) 165.24
IUPAC-namn N,N-dimethyl-2-phenoxyethanamine
LEDER CN(C)CCOc1ccccc1
Specifikationer
Kemiskt namn eller material N,N-Dimethyl-2-phenoxyethanamine
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C10H15NO/c1-11(2)8-9-12-10-6-4-3-5-7-10/h3-7H,8-9H2,1-2H3
Formel vikt 165.1154
Procent renhet >95%