Läs mer
N,N-Dimethyl-2-phenylethanamine, TRC
Produktkod. 30010531
Change view
Kvantitet:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010531 10 mL
17920775 1 mL
17904985 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010531 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB41303010ML
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1126-71-2
Molekylformel C10H15N
Molekylvikt (g/mol) 149.23
Synonym N,N-Dimethylbenzeneethanamine, Dimethyl(2-phenylethyl)amine, Dimethylphenethylamine, N,N-Dimethyl-beta-phenethylamine, N,N-Dimethylphenethylamine;N-Phenethyldimethylamine, NSC 1849;
IUPAC-namn N,N-dimethyl-2-phenylethanamine
LEDER CN(C)CCc1ccccc1
Specifikationer
Kemiskt namn eller material N,N-Dimethyl-2-phenylethanamine
Kvantitet 10 mL
InChI formel InChI=1S/C10H15N/c1-11(2)9-8-10-6-4-3-5-7-10/h3-7H,8-9H2,1-2H3
Formel vikt 149.1204