missing translation for 'onlineSavingsMsg'
Läs mer

N,N-Dimethyl-5-[[[2-[[1-(methylthio)-2-nitroethenyl]amino]ethyl]thio]methyl]-2-furanmethanamine, TRC

Product Code. 30379844
Change view
Click to view available options
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30379844 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379844 Supplier Toronto Research Chemicals Supplier No. TRCD47358050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-[2-(5-Dimethylaminomethyl-2-furanyl)methylthio]ethylamino]-1-methylthio-2-nitroethene
CAS 72115-14-1
Molekylformel C13 H21 N3 O3 S2
Kvantitet 50 mg
Synonym 2-Furanmethanamine, N,N-dimethyl-5-[[[2-[[1-(methylthio)-2-nitroethenyl]amino]ethyl]thio]methyl]-, N-[2-[[5-[(dimethylamino)methyl]-2-furyl]methylsulfanyl]ethyl]-1-methylsulfanyl-2-nitro-ethenamine, 1-[2-(5-Dimethylaminomethyl-2-furanyl)methylthio]ethylamino]-1-methylthio-2-nitroethene
InChI formel InChI=1S/C13H21N3O3S2/c1-15(2)8-11-4-5-12(19-11)10-21-7-6-14-13(20-3)9-16(17)18/h4-5,9,14H,6-8,10H2,1-3H3
LEDER CSC(=C[N+](=O)[O-])NCCSCc1oc(CN(C)C)cc1
IUPAC-namn N-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-methylsulfanyl-2-nitroethenamine
Molekylvikt (g/mol) 331.45
Formel vikt 331.1024
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.