Läs mer
N,N-Dimethyl-carbamic Acid C,C'-[5-(2-Bromoacetyl)-1,3-phenylene] Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N,N-Dimethyl-carbamic Acid C,C'-[5-(2-Bromoacetyl)-1,3-phenylene] Ester |
| CAS | 81732-49-2 |
| Molekylformel | C14H17BrN2O5 |
| Kvantitet | 250 mg |
| Synonym | N,N-Dimethyl-carbamic Acid 5-(Bromoacetyl)-1,3-phenylene Ester, 5-(2-Bromoacetyl)-1,3-phenylene Bis(dimethylcarbamate) |
| InChI formel | InChI=1S/C14H17BrN2O5/c1-16(2)13(19)21-10-5-9(12(18)8-15)6-11(7-10)22-14(20)17(3)4/h5-7H,8H2,1-4H3 |
| LEDER | O=C(N(C)C)OC1=CC(C(CBr)=O)=CC(OC(N(C)C)=O)=C1 |
| IUPAC-namn | 5-(2-bromoacetyl)-1,3-phenylene bis(dimethylcarbamate) |
| Molekylvikt (g/mol) | 373.2 |
| Formel vikt | 372.03 |