Läs mer
N,N-Dimethyl-L-valine, TRC
Produktkod. 30373847
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30373847 5 g
1 options
Produktkod. 30373847 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4796455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N-Dimethyl-L-valine
CAS 2812-32-0
Molekylformel C7 H15 N O2
Kvantitet 5 g
Synonym N,N-Dimethylvaline, (S)-2-(Dimethylamino)-3-methylbutanoic Acid
InChI formel InChI=1S/C7H15NO2/c1-5(2)6(7(9)10)8(3)4/h5-6H,1-4H3,(H,9,10)/t6-/m0/s1
LEDER CC(C)[C@H](N(C)C)C(=O)O
IUPAC-namn (2S)-2-(dimethylamino)-3-methyl-butanoic acid
Molekylvikt (g/mol) 145.2
Formel vikt 145.1103