Läs mer
N,N'-Dimethyl-N,N'-bis(mercaptoacetyl)hydrazine(>90%), TRC
Produktkod. 30366961
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30366961 50 mg
1 options
Produktkod. 30366961 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD13567050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-Dimethyl-N,N'-bis(mercaptoacetyl)hydrazine
CAS 131760-67-3
Molekylformel C6 H12 N2 O2 S2
Kvantitet 50 mg
Synonym 2-Mercaptoacetic Acid 2-(2-mercaptoacetyl)-1,2-dimethylhydrazide, Mercapto-2-(mercaptoacetyl)-1,2-dimethylhydrazide Acetic Acid
InChI formel InChI=1S/C6H12N2O2S2/c1-7(5(9)3-11)8(2)6(10)4-12/h11-12H,3-4H2,1-2H3
LEDER CN(N(C)C(=O)CS)C(=O)CS
IUPAC-namn N,N'-dimethyl-2-sulfanyl-N'-(2-sulfanylacetyl)acetohydrazide
Molekylvikt (g/mol) 208.302
Formel vikt 208.034