Läs mer
N,N-Dimethyl-o-toluidine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N,N-Dimethyl-o-toluidine |
| CAS | 609-72-3 |
| Molekylformel | C9H13N |
| Kvantitet | 250 g |
| Synonym | 2,N,N-Trimethylaniline, 2-Methyl-N,N-dimethylaniline, Dimethyl-o-toluidine, N,N,2-Trimethylaniline, N,N-Dimethyl-2-methylaniline, N,N-Dimethyl-o-methylaniline, NSC 1784, o-Methyl-N,N-dimethylaniline, o-Methyldimethylaniline |
| InChI formel | InChI=1S/C9H13N/c1-8-6-4-5-7-9(8)10(2)3/h4-7H,1-3H3 |
| LEDER | CN(C)c1ccccc1C |
| Molekylvikt (g/mol) | 135.21 |
| Formel vikt | 135.1048 |