Learn More
N,N-Dimethylacetamide, ≥99%, Spectrophotometric Grade, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
Description
- Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
Specifications
Melting Point | -20°C |
Linear Formula | CH3CON(CH3)2 |
Beilstein | 1737614 |
Formula Weight | 87.12g/mol |
Percent Purity | ≥99% |
Grade | Spectrophotometric |
Density | 0.937g/mL at 25°C |
Boiling Point | 164°C to 166°C |
Quantity | 1 L |
Explosion Limit | 1.8% (100°F), 11.5% (160°F) |