missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5391018
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
127-19-5
C4H9NO
87.12
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CN(C)C(C)=O
This item is not returnable. View return policy
127-19-5
C4H9NO
87.12
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CN(C)C(C)=O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 87.12
Grade BAKER ANALYZED™ Reagent
Quantity 500 mL
Flash Point 70.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.94kg
Chemical Name or Material N,N-Dimethylacetamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'