missing translation for 'onlineSavingsMsg'
Läs mer

N,N-Dimethylethylenediamine, Sigma-Aldrich™

Produktkod. 32238306
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
22 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
32238306 22 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 32238306 Leverantör Sigma Aldrich Leverantörsnummer 8037799022
 Begränsad artikel
Beräknad leverans: 26-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

For synthesis

N,N-Dimethylethylenediamine (DMEDA) is used as a bidentate ligand in coordination chemistry due to the presence of two nitrogen atoms, one with lone pairs that can coordinate to a metal center. It is used as a ligand and base in Suzuki-Miyaura and Sonogashira coupling reactions, enhancing palladium catalyst stability, and deprotonating alkynes. It acts as a nucleophile and a base, promoting reactions involving proton transfer or nucleophilic attack. It is commonly used in amination reactions to synthesize substituted amines and in epoxide ring openings to form amino alcohols. Additionally, DMEDA serves as a chelating agent, forming stable complexes with transition metals. It acts as a secondary amine in reductive amination to synthesize tertiary amines.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material N,N-Dimethylethylenediamine
CAS 108-00-9
Kokområde 107°C to 109°C/1013 hPa
Smältpunkt <-70°C
Densitet 0.81 g/cm³ at 20°C
Flampunkt 11.7°C (closed cup)
Molekylformel NH2C2H4N(CH3)2
Kvantitet 22 kg
Synonym N,N-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, N,N-Dimethylethylenediamine
Molekylvikt (g/mol) 88.15
Kvalitet Not Available
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.