Learn More
N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Description
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | °C to 61°C |
Color | Colorless |
Assay Percent Range | 99.8% |
UN Number | 2265 |
Beilstein | 605365 |
Solubility Information | Miscible with water and most organic solvents. |
Formula Weight | 73.09 |
Density | 0.944 g/cm3 at 20°C |
Boiling Point | 153°C |
Quantity | 100 mL |
Show More |
RUO – Research Use Only