Läs mer
N,N-Dimethylmelamine Metformin Impurity C, TRC
Produktkod. 30137850
Change view
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137850 100 mg
17960585 1 g
17943495 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137850 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD474600100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1985-46-2
Molekylformel C5 H10 N6
Molekylvikt (g/mol) 154.17
Synonym N,N-Dimethyl-1,3,5-triazine-2,4,6-triamine, Metformin Hydrochloride Imp. C (EP)
IUPAC-namn 2-N,2-N-dimethyl-1,3,5-triazine-2,4,6-triamine
LEDER CN(C)c1nc(N)nc(N)n1
Specifikationer
Kemiskt namn eller material N,N-Dimethyl-1,3,5-triazine-2,4,6-triamine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C5H10N6/c1-11(2)5-9-3(6)8-4(7)10-5/h1-2H3,(H4,6,7,8,9,10)
Formel vikt 154.0967
Procent renhet >95%