Läs mer
N,N-Dimethylmethanamine Oxide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N,N-Dimethylmethanamine Oxide |
| CAS | 1184-78-7 |
| Molekylformel | C3H9NO |
| Kvantitet | 100 mg |
| Synonym | Trimethylamine, N-oxide (6CI, 8CI), N,N-Dimethylmethanamine N-oxide, NSC 408426, TMAO, Trimethylamine oxide, Triox |
| InChI formel | InChI=1S/C3H9NO/c1-4(2,3)5/h1-3H3 |
| LEDER | C[N+](C)(C)[O-] |
| IUPAC-namn | N,N-dimethylmethanamine oxide |
| Molekylvikt (g/mol) | 75.11 |
| Formel vikt | 75.0684 |