Learn More
N,N-Dimethylmethyleneiminium iodide, 97%, Thermo Scientific Chemicals
Catalog Number p-126378
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
33797-51-2
C3H8IN
185.01
MFCD00011810
VVDUZZGYBOWDSQ-UHFFFAOYSA-M
eschenmoser's salt, n,n-dimethylmethyleneiminium iodide, n-methyl-n-methylenemethanaminium iodide, dimethyl methylidene azanium iodide, n,n-dimethyl methyleneammonium iodide, dimethyl methylene ammonium iodide, methanaminium, n-methyl-n-methylene-, iodide, eschenmoser inverted exclamation mark s salt, eschenmoser salt, dimethyl methylene ammounium iodide
2724133
dimethyl(methylidene)azanium;iodide
C[N+](=C)C.[I-]
33797-51-2
C3H8IN
185.01
MFCD00011810
VVDUZZGYBOWDSQ-UHFFFAOYSA-M
eschenmoser's salt, n,n-dimethylmethyleneiminium iodide, n-methyl-n-methylenemethanaminium iodide, dimethyl methylidene azanium iodide, n,n-dimethyl methyleneammonium iodide, dimethyl methylene ammonium iodide, methanaminium, n-methyl-n-methylene-, iodide, eschenmoser inverted exclamation mark s salt, eschenmoser salt, dimethyl methylene ammounium iodide
2724133
dimethyl(methylidene)azanium;iodide
C[N+](=C)C.[I-]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 219°C
Color Cream to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2C=N(CH3)2I
Quantity 10 g
Beilstein 04, IV, 153
Fieser 04,186; 08,194; 10,160; 13,121
Merck Index 15, 3275
Formula Weight 185.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Dimethylmethyleneiminium iodide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Dimethylmethyleneiminium iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.